$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
BUFFER TRIS (D11) (DOCD2)3CND2 98% 5 G
Catalog # 103062-126
Supplier:
Cambridge Isotope Labs Inc
Expand image
BUFFER TRIS (D11) (DOCD2)3CND2 98% 5 G
Catalog # 103062-126
Supplier:
Cambridge Isotope Labs Inc
Supplier Number:
DLM-1814-5