Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SI-TRIMERCAPTOTRIAZINE RESIN 40-62 U 1KG
Catalog # 103062-092
Supplier:
Supra Sciences
SDS
Expand image
SI-TRIMERCAPTOTRIAZINE RESIN 40-62 U 1KG
Catalog # 103062-092
Supplier:
Supra Sciences
Supplier Number:
SPSI22-10
SDS
Related Information
Documentation
SDS
ghs_SPSI22
Documentation