$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
DIETHANOLAMINE-FINE RESIN DEAM FUNC 10G
103062-006
:
Supra Sciences
DIETHANOLAMINE-FINE RESIN DEAM FUNC 10G
103062-006
:
Supra Sciences
:
SPMP23-10
ghs_SPMP23