Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RESIN PS-AMINOMETHYL 100-200 MESH 1000 G
Catalog # 103061-852
Supplier:
Supra Sciences
SDS
Expand image
RESIN PS-AMINOMETHYL 100-200 MESH 1000 G
Catalog # 103061-852
Supplier:
Supra Sciences
Supplier Number:
SPPS37-1KG
SDS
Related Information
Documentation
Datasheet
Aminomethyl.pdf
SDS
ghs_SPPS37
Documentation