Zenix™ SEC Columns, Sepax Technologies
Analytical, Zenix™ SEC-80, 80 Å, 3 µm, 2.10 mm, 300 mm, Neutral, hydrophilic film bonded silica
Catalog # 103061-448
Marketsource
Supplier: Sepax Technologies
: 213080-2130
Zenix™ SEC Columns, Sepax Technologies
Catalog # 103061-448
Supplier: Sepax Technologies
Marketsource
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Zenix™ SEC phase, developed based on innovative surface coating technology, comprises of uniform, hydrophilic, and neutral nanometer thick film chemically bonded to high purity and mechanically stabilized silica.
- Highest separation efficiency and resolution
- High loading capacity
- High stability over low and high concentration salt
- Lot-to-lot reproducibility
- Aqueous and organic mobile phase compatibility
- High protein recovery with intact biological activity
- Negligible non-specific interactions
- Column packed with PEEK tubing
Zenix™ uses lay-down monolayer on porous silica offers ideal phase chemistry for hydrophobic samples like insulin, membrane protein monoclonal antibodies derivatized with polymer branches (polypeptide or PEG) separation.
Ideal for separation and analysis of biological molecules such as proteins, nucleic acids, oligonucleotides, peptides, virus and natural polymers such as polysaccharides, synthetic polymers and nanomaterials such as nanoparticles.
Specifications
- Column Format:Analytical
- Ø int.:2.10 mm
- Particle Size:3 µm
- Pore size:80 Å
- Length:300 mm
- Packing material:Neutral, hydrophilic film bonded silica
- Phase:Zenix™ SEC-80
- Cat. No.:103061-448
- pH Range:2 - 8.5
- Temperature Limit:80 °C



