Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
5-BR-4-CL-3-INDOXYL-A-L-ARABINOFURA 0.1G
Catalog # 103052-330
Supplier:
Biosynth International Inc
SDS
Expand image
5-BR-4-CL-3-INDOXYL-A-L-ARABINOFURA 0.1G
Catalog # 103052-330
Supplier:
Biosynth International Inc
Supplier Number:
EB04909-0.1G
SDS
Related Information
Documentation
SDS
ghs_B-7044.pdf
Documentation