Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
5-METHOXY-DL-TRYPTOPHAN 28052-84-8 10G
Catalog # 103052-218
Supplier:
Biosynth International Inc
SDS
Expand image
5-METHOXY-DL-TRYPTOPHAN 28052-84-8 10G
Catalog # 103052-218
Supplier:
Biosynth International Inc
Supplier Number:
FM30488-10G
SDS
Related Information
Documentation
SDS
ghs_M-3570.pdf
Documentation