Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
5-BROMO-DL-TRYPTOPHAN 6548-09-0 5G
Catalog # 103052-144
Supplier:
Biosynth International Inc
SDS
Expand image
5-BROMO-DL-TRYPTOPHAN 6548-09-0 5G
Catalog # 103052-144
Supplier:
Biosynth International Inc
Supplier Number:
FB52324-100MG
SDS
Related Information
Documentation
SDS
ghs_B-9100.pdf
Documentation