$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
5-BROMO-4-CHLORO-3-INDOXYL-1-ACETATE 5G
Catalog # 103052-140
Supplier:
Biosynth International Inc
SDS
Expand image
5-BROMO-4-CHLORO-3-INDOXYL-1-ACETATE 5G
Catalog # 103052-140
Supplier:
Biosynth International Inc
Supplier Number:
EB56757-5G
SDS
Related Information
Documentation
SDS
ghs_B-6000.pdf
Documentation