$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
N-DODECYL-BETA-D-MALTOSIDE C24H46O11 25G
103051-762
:
Biosynth International Inc
N-DODECYL-BETA-D-MALTOSIDE C24H46O11 25G
103051-762
:
Biosynth International Inc
:
DD06199-25G
ghs_D-8823.pdf