Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
L-5-HYDROXYTRYPTOPHAN DIHYDRATE 250 G
Catalog # 103051-526
Supplier:
Biosynth International Inc
SDS
Expand image
L-5-HYDROXYTRYPTOPHAN DIHYDRATE 250 G
Catalog # 103051-526
Supplier:
Biosynth International Inc
Supplier Number:
FH172368-10G
SDS
Related Information
Documentation
SDS
ghs_H-8330.pdf
Documentation