Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
D-MELEZITOSE CAS NUMBER 10030-67-8 1000G
Catalog # 103051-424
Supplier:
Biosynth International Inc
SDS
Expand image
D-MELEZITOSE CAS NUMBER 10030-67-8 1000G
Catalog # 103051-424
Supplier:
Biosynth International Inc
Supplier Number:
OM00386-1000G
SDS
Related Information
Documentation
SDS
ghs_M-1520.pdf
Documentation