Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
N-METHYLINDOXYL-B -D-GALACTO 0.1G
103051-410
:
Biosynth International Inc
N-METHYLINDOXYL-B -D-GALACTO 0.1G
103051-410
:
Biosynth International Inc
:
EM04984-0.1G
ghs_M-4100