$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
4-NITROPHENYL-BETA-D-XYLOPYRANOSIDE 10G
103051-222
:
Biosynth International Inc
4-NITROPHENYL-BETA-D-XYLOPYRANOSIDE 10G
103051-222
:
Biosynth International Inc
:
EN03230-10G
ghs_N-5300.pdf