$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
CALIBRATION POUCHES PH 7.00 CS20
MSPP-MW7
:
PENTAIR AQUATIC ECO-SYSTEMS INC MS
CALIBRATION POUCHES PH 7.00 CS20
MSPP-MW7
:
PENTAIR AQUATIC ECO-SYSTEMS INC MS
:
MW7