Order Entry
Puerto Rico
ContactUsLinkComponent
DRYING/HEATING CHAMBER 230V STANDARD 56L
Catalog # 75783-600
Supplier:  Binder
undefined
DRYING/HEATING CHAMBER 230V STANDARD 56L
Catalog # 75783-600
Supplier:  Binder
Supplier Number:  9010-0303