Order Entry
Puerto Rico
ContactUsLinkComponent
DRYING/HEATING CHAMBER 230V FORCED 115L
Catalog # 75783-588
Supplier:  Binder
undefined
DRYING/HEATING CHAMBER 230V FORCED 115L
Catalog # 75783-588
Supplier:  Binder
Supplier Number:  9010-0293