TWD Barcoded Autosampler Vials
Cap only, black
Catalog # 75780-320
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
CAP SCREW CLEAR PP 9MM SILICONE PK100 Catalog #AG5191-8151
Product AG5191-8151 is similar to 75780-320 but manufacturer and specifications may vary.
Product Details & Documents
TWD Barcoded Autosampler Vials with 2D barcode.
- Vials feature writing patch and graduations
- Glass construction
- Screw-top 9-425 GPI thread finish
TWD barcoded autosampler kits are a cost-effective option if you're looking for a easy, hassle-free option to track your samples your samples.
Delivery information: Convenient Kit includes black PP cap and red rubber/PTFE Septa; available as vial and cap kit or cap only.
Specifications
- Description:Cap only, black