Corning® CoolCell® Containers, Corning
Product Details & Documents
CoolCell® alcohol-free cell freezing containers ensure standardized controlled-rate −1 °C/minute cell freezing in a −80 °C freezer, without alcohol or any fluids.
- No alcohol. no fluids, no pre-cooling
- No variability, all vials have uniform freeze rate
- Radially symmetric design ensures vial consistency
- No on-going cost, no alcohol purchase or disposal
- No stuck lids, ergonomic lid comes off easily when frozen
- Not cold to the touch when removing from the −80 ˚C freezer
- Quick re-use time, ready to use again after five minutes
Proven for use with a variety of cell types including stem cells, primary cells, PBMC cell lines, insect cells, yeast and others. The patent-pending CoolCell® technology utilizes a thermo-conductive alloy core and highly-insulative outer material to control the rate of heat removal and provide reproducible cell cryopreservation. CoolCell units are easy to use and deliver comparable results to expensive programmable freezers.
CoolCell® LX
For 12 standard 1,0 to 2,0 ml cryogenic vials, 1,0 ml fill per vial. Radially symmetric for uniform vial freezing. Numbered wells for easy sample identification. Beveled lid for secure gripping and easy opening. Exposed vial tops when lid is open for quick, organized removal of frozen samples.
CoolCell® FTS30
For 30 standard 1,0 to 2,0 ml cryogenic vials, 1,0 ml fill per vial. Controlled micro-convection for uniform freezing of 30 vials. Removable vial tray for one-step transfer of samples into and out of freezing chamber."
CoolCell® 5 mL LX
For 12 standard 3,5 to 5,0 ml fill cryogenic vials, 3,5 to 5,0 fill per vial. Radially symmetric for uniform vial freezing. Numbered wells for easy sample identification. Beveled lid for secure gripping and easy opening. Exposed vial tops when lid is open for quick, organized removal of frozen samples.
CoolCell® SV2 and CoolCell® SV10
For 12 standard 2,0 ml injectable ampules, 1,0 ml fill per ampule (SV2). For 6 standard 10,0 ml injectable ampules, 5,0 ml fill per ampule (SV10). Radially symmetric for uniform freezing of injectable ampules. Easy open lid. Exposed vial tops when lid is open for quick, organized removal of frozen samples.
CoolCell SV10 Stem Cell Cryopreservation System
includes the XT Starter Benchtop Cooler, Corning® CoolRack SV10 and CoolCell SV10 Cell Freezing Container for 6 injectable vials.
CoolCell® Filler Vials
To ensure cell freezing rate consistency and uniform results when using CoolCell containers, insert a CoolCell Filler Vial into empty wells when freezing less than a full load. Suitable for repeated use and compatible with CoolCell LX, CoolCell FTS30 and CoolCell 5 ml LX containers.
CoolCell® FTS30 Vial Module
FTS30 Vial Module is a holder for 30×1,0 ml or 2,0 ml cryogenic vials that allows one-step insertion and removal of all 30 vials at once. Fits into a standard 5,0×5,0×2,0" cryostorage box. Compatible with dry ice and liquid nitrogen.
Documents
Specifications
- No. of Holes:6
- Color:Purple
- Description:CoolCell® SV10 Alchol-free Freezing Container, Exposed Vial Tops
- Dimensions:12.1×9.8 cm
- For vials:10 ml serum vials
- Well diameter:23.6 mm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Removable Cryogenic vial Module for CoolCell FTS30, 3/Pk.Removable Cryogenic vial Module for CoolCell FTS30, 3/Pk.75779-782Log in for availabilityCase of 10 - $153.00
Product Information
- Catalog #AvailabilityPrice
- CoolCell® 2 mL Filler Vials for Use with CoolCell LX and CoolCell FTS30 ContainersCoolCell® 2 mL Filler Vials for Use with CoolCell LX and CoolCell FTS30 Containers75779-778Log in for availabilityCase of 6 (2ml) - $36.20
Product Information
- Catalog #AvailabilityPrice
- Cryogenic Vial Grippers, Pack of 5, Assorted ColorsCryogenic Vial Grippers, Pack of 5, Assorted Colors75779-848Log in for availabilityCase of 5 - $70.80
Product Information
- Catalog #AvailabilityPrice
- CoolCell®5 mL Filler Vials for Use with CoolCell 5 mL LX ContainerCoolCell®5 mL Filler Vials for Use with CoolCell 5 mL LX Container75779-780Log in for availabilityCase of 6 (5ml) - $36.20
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



