Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Rigid-Lock QUICKBERM®, Justrite®

EnGuard™V, modified PVC coated fabric, 66L x 66W x 30.5H cm (26 x 26 x 12"), 243.8W x 243.8D x 30.5H cm (96 x 96 x 12"), 264.2L x 264.2W x 30.5H cm (104 x 104 x 12"), 1798 L (475 gal.)
Catalog # 10861-140
Supplier:   Justrite
 :  28518
Rigid-Lock QUICKBERM®, Justrite®
Catalog # 10861-140
Supplier:  Justrite
Rigid-Lock QUICKBERM®, Justrite®
Rigid-Lock QUICKBERM®, Justrite®

EnGuard™V, modified PVC coated fabric, 66L x 66W x 30.5H cm (26 x 26 x 12"), 243.8W x 243.8D x 30.5H cm (96 x 96 x 12"), 264.2L x 264.2W x 30.5H cm (104 x 104 x 12"), 1798 L (475 gal.)

Price
$1,871.56
Log in for availability
Each (475US Gal)
Product Details & Documents

Ergonomic design provides 100% inside berm accessibility with an unobstructed side-to-side floor space for vehicle tires, foot traffic, and cleaning.

  • Single-piece construction - patented Rigid-Lock wall-support system integral to berm provides unobstructed work space
  • Quick and easy setup - no assembly required
  • Durable EnGuard™V, modified PVC coated fabric withstands long-term UV exposure

Exceptionally strong structural integrity includes a reinforced CriticalCorner™ design featuring wraparound diagonal radio frequency welds to prevent billowing, unwrapping, or sagging of the corners during full containment for fluid-tight reliability. Sidewalls lower quickly for easy drive-in and out. Portable design folds down quickly for storage and transport.

The built-in Rigid-Lock wall supports maximize usable space inside the berm and greatly reduce any tripping hazard on the berm perimeter. The device only folds down one way, inward towards the inside of the berm. The one-piece design folds down flat and remains integral to the sidewall. Supports resist extreme temperatures (-50 to 160ºF). When in the down position, they can withstand vehicle weights in excess of 11,000 lbs (5,000 kg) at each tire

Specifications
  • Material:
    EnGuard™V, modified PVC coated fabric
  • Interior dimensions:
    243.8W x 243.8D x 30.5H cm (96 x 96 x 12")
  • Ext. dimensions:
    264.2L x 264.2W x 30.5H cm (104 x 104 x 12")
  • Sump capacity:
    1798 L (475 gal.)
  • Folded size:
    66L x 66W x 30.5H cm (26 x 26 x 12")