Various Replacement Parts, Sciencix
Indicator Rod Kit, 510, 515, 600, 610, 1515, 1525, 1525 Micro, 1525EF, WATE, WAT069583
Catalog # 103011-966
Marketsource
Supplier: SCIENCIX
: CTS-6958
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Indicator Rod Kit
- OEM manufacturer:WATE
- OEM equivalent:WAT069583
- Model:510, 515, 600, 610, 1515, 1525, 1525 Micro, 1525EF