Various Replacement Parts, Sciencix
Solvent Bottle Filter, ACQUITY I-Class BSM, ACQUITY I-Class SM-FTN, ACQUITY UPLC I2V BSM, ACQUITY UPLC BSM, WATE, 700003616
Catalog # 103011-948
Marketsource
Supplier: SCIENCIX
: CTS-5531-07
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Solvent Bottle Filter
- OEM manufacturer:WATE
- OEM equivalent:700003616
- Model:ACQUITY I-Class BSM, ACQUITY I-Class SM-FTN, ACQUITY UPLC I2V BSM, ACQUITY UPLC BSM