Various Replacement Parts, Sciencix
Precolumn Filter Insert, 2 µm, 600, 717, 2690, 2690D, 2695, 2695D, 2790, 2795, 2796, Alliance, LC Module 1, WATE, WAT088084
Catalog # 103011-902
Marketsource
Supplier: SCIENCIX
: CTS-10919
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Precolumn Filter Insert, 2 µm
- OEM manufacturer:WATE
- OEM equivalent:WAT088084
- Model:600, 717, 2690, 2690D, 2695, 2695D, 2790, 2795, 2796, Alliance, LC Module 1