Various Replacement Parts, Sciencix
PEEK Ferrule, 1/16", ACQUITY UPLC High Temperature Column Heater, ACQUITY UPLC Column Heater Module, ACQUITY UPLC Column Heater/Cooler, ACQUITY UPLC Column Manager, WATE, 700003114
Catalog # 103011-788
Marketsource
Supplier: SCIENCIX
Supplier #: 11-2414
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:PEEK Ferrule, 1/16"
- OEM manufacturer:WATE
- OEM equivalent:700003114
- Model:ACQUITY UPLC High Temperature Column Heater, ACQUITY UPLC Column Heater Module, ACQUITY UPLC Column Heater/Cooler, ACQUITY UPLC Column Manager