Orders LinkContactUsLinkComponent
Various Replacement Parts, Sciencix
Needle Seal, Vespel®, LC-2010, SIL-10ADvp, SIL-10AXL, SHIM, 228-33355-04
Catalog # 103011-732
Marketsource
Supplier:   SCIENCIX
 :  CTS-10686
Various Replacement Parts, Sciencix
Catalog # 103011-732
Supplier:  SCIENCIX
Marketsource
Various Replacement Parts, Sciencix
Various Replacement Parts, Sciencix
Needle Seal, Vespel®, LC-2010, SIL-10ADvp, SIL-10AXL, SHIM, 228-33355-04
Price
$79.98
Log in for availability
Each
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
  • Ensures your system is working efficiently
  • Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
  • Description:
    Needle Seal, Vespel®
  • OEM manufacturer:
    SHIM
  • OEM equivalent:
    228-33355-04
  • Model:
    LC-2010, SIL-10ADvp, SIL-10AXL