Various Replacement Parts, Sciencix
Diaphragm, PTFE, LC-10AD, LC-10ADvp, LC-20AD/AB, LC-20ADXR, LC-30ADSF, LC-2010, LC-2010 HT, SHIM, 228-32784-91
Catalog # 103011-706
Marketsource
Supplier: SCIENCIX
: 11-2891
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Diaphragm, PTFE
- OEM manufacturer:SHIM
- OEM equivalent:228-32784-91
- Model:LC-10AD, LC-10ADvp, LC-20AD/AB, LC-20ADXR, LC-30ADSF, LC-2010, LC-2010 HT