Various Replacement Parts, Sciencix
Fingertight One Piece 10 – 32 PEEK Male Nut/Ferrule for 1/16" OD Tubing, (Agilent) 1050, 1090, 1100, 1200, 1220
(PerkinElmer) 200 Series, 1,2,3,3B, 4,10, 250, 400, 410, 620, Int. 4000, Agilent
PerkinElmer, 5063-6591
ED020005
(PerkinElmer) 200 Series, 1,2,3,3B, 4,10, 250, 400, 410, 620, Int. 4000, Agilent
PerkinElmer, 5063-6591
ED020005
Catalog # 103011-704
Marketsource
Supplier: SCIENCIX
Supplier #: 11-1240
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Fingertight One Piece 10 – 32 PEEK Male Nut/Ferrule for 1/16" OD Tubing
- OEM manufacturer:Agilent
PerkinElmer - OEM equivalent:5063-6591
ED020005 - Model:(Agilent) 1050, 1090, 1100, 1200, 1220
(PerkinElmer) 200 Series, 1,2,3,3B, 4,10, 250, 400, 410, 620, Int. 4000