Various Replacement Parts, Sciencix
Capillary, 0.17ID×400L mm with Fittings, 1100, 1200, 1260, G1312, AGLS, G1312-87303
Catalog # 103011-698
Marketsource
Supplier: SCIENCIX
Supplier #: CTS-A11459
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Capillary, 0.17ID×400L mm with Fittings
- OEM manufacturer:AGLS
- OEM equivalent:G1312-87303
- Model:1100, 1200, 1260, G1312