Various Replacement Parts, Sciencix
Seal Cap Assembly, 1050, 1100, 1200, 1220, 1260, AGLS, 5067-4728
Catalog # 103011-694
Marketsource
Supplier: SCIENCIX
: CTS-A11454
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Seal Cap Assembly
- OEM manufacturer:AGLS
- OEM equivalent:5067-4728
- Model:1050, 1100, 1200, 1220, 1260