Orders LinkContactUsLinkComponent
Various Replacement Parts, Sciencix
Capillary, 0.17ID×280L mm, 1100, 1200, 1260, AGLS, 5021-1818
Catalog # 103011-684
Marketsource
Supplier:   SCIENCIX
 :  CTS-11458
Various Replacement Parts, Sciencix
Catalog # 103011-684
Supplier:  SCIENCIX
Marketsource
Various Replacement Parts, Sciencix
Various Replacement Parts, Sciencix
Capillary, 0.17ID×280L mm, 1100, 1200, 1260, AGLS, 5021-1818
Price
$49.88
Log in for availability
Each
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
  • Ensures your system is working efficiently
  • Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
  • Description:
    Capillary, 0.17ID×280L mm
  • OEM manufacturer:
    AGLS
  • OEM equivalent:
    5021-1818
  • Model:
    1100, 1200, 1260