Various Replacement Parts, Sciencix
Glass Solvent Filter, 1050, 1090, 1100, 1200, 1220, 1260, 1290, AGLS, 5041-2168
Catalog # 103011-654
Marketsource
Supplier: SCIENCIX
Supplier #: CTS-10843
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Glass Solvent Filter
- OEM manufacturer:AGLS
- OEM equivalent:5041-2168
- Model:1050, 1090, 1100, 1200, 1220, 1260, 1290