Various Replacement Parts, Sciencix
Compression Screw, Stainless Steel, 710, 712, 715, 717, 2690, 2690D, 2695, 2695D, 2790, 2795, WATE, WAT025313
Catalog # 103011-634
Marketsource
Supplier: SCIENCIX
: CTS-5313
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Compression Screw, Stainless Steel
- OEM manufacturer:WATE
- OEM equivalent:WAT025313
- Model:710, 712, 715, 717, 2690, 2690D, 2695, 2695D, 2790, 2795