Various Replacement Parts, Sciencix
Filter, Air, Chassis, ACQUITY UPLC Sample Mgr, nanoACQUITY UPLC Sample Mgr, WATE, 401000501
Catalog # 103011-594
Marketsource
Supplier: SCIENCIX
: CTS-11150
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Ensures your system is working efficiently
- Brand new replacement parts
Choose from various batteries, filters, frits, and other replacement parts to ensure your Sciencix chromatography system is working efficiently.
Specifications
- Description:Filter, Air, Chassis
- OEM manufacturer:WATE
- OEM equivalent:401000501
- Model:ACQUITY UPLC Sample Mgr, nanoACQUITY UPLC Sample Mgr