Protocol Station, Restek
Product Details & Documents
The protocol station is designed for convenient wall mounting of high-purity gas regulators.
- Fabricated in stainless steel or chrome-plated brass
- Provides ease of use for high-purity gas regulators
Wall mounting provides ease of use, prevents gas regulator damage, and improves safety. Either chrome-plated brass or 316 stainless steel option is complete with a 3-foot, flexible, all-stainless-steel pigtail with armor casing. The CGA connection on the pigtail has an integral check valve in the gland to prevent contamination during cylinder changeout.
Ordering information: Pressure regulator not included. Order separately.
Specifications
- Description:Protocol Station*
- Material:Brass, chrome plated
- Includes:CGA 590 (Air)