Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Substance P

Substance P

5 mg
Catalog # 102996-506
Marketsource
Supplier:   Anaspec
Supplier #:  AS-24280
Substance P
Catalog # 102996-506
Supplier:  Anaspec
Marketsource
CAS Number:  
Substance P

5 mg

CAS Number:  
Price
$160.77
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Substance P (SP) is an important neuropeptide belonging to the tachykinin family. It acts as a neurotransmitter and neuromodulator. It is closely related to neurokinin A, both originating from the same precursor preprotachykinin A. It is released from the terminals of sensory nerves and is involved in inflammation and pain processes.
Sequence:RPKPQQFFGLM-NH2
MW:1347.7 Da
% peak area by HPLC:95
Storage condition:-20° C

Specifications

Specifications

  • Size:
    5 mg
  • Storage Conditions:
    –20 °C
  • Cat. No.:
    102996-506
  • Molecular Weight:
    1347.7 Da
  • Purity:
    95%
  • Protein Synonyms:
    Substance P neuropeptide
  • Protein/Peptide Name:
    Substance P
  • Conjugation:
    Unconjugated
  • Sequence:
    RPKPQQFFGLM-NH2

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.