Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
KIT BLOOD DNA MINI-IPC16 INNUPREP 96 RXN
Catalog # 10852-478
Supplier:
Analytik Jena US
SDS
Expand image
KIT BLOOD DNA MINI-IPC16 INNUPREP 96 RXN
Catalog # 10852-478
Supplier:
Analytik Jena US
Supplier Number:
845IPP-1016096
SDS
Related Information
Documentation
SDS
ghs_845IPP-1016096
Documentation