Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
INNUPREP BLOOD DNA MIDI KIT-IPC16 96RXN
Catalog # 10852-442
Supplier:
Analytik Jena US
SDS
Expand image
INNUPREP BLOOD DNA MIDI KIT-IPC16 96RXN
Catalog # 10852-442
Supplier:
Analytik Jena US
Supplier Number:
845-IPS-1216096
SDS
Related Information
Documentation
SDS
ghs_845IPS-1216096.pdf
Documentation