Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
KIT FOOD DNA INNUPREP 480 REACTIONS PLT
Catalog # 10852-434
Supplier:
Analytik Jena US
SDS
Expand image
KIT FOOD DNA INNUPREP 480 REACTIONS PLT
Catalog # 10852-434
Supplier:
Analytik Jena US
Supplier Number:
845-IPP-5716480
SDS
Related Information
Documentation
SDS
ghs_845IPP-5716480.pdf
Documentation