Danusertib ≥95%
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Aurora kinases are key regulators of protein phosphorylation during mitosis, and these serine/threonine kinases interact with other cellular proteins to help control chromosome assembly and segregation during mitosis. Aurora kinases are often highly expressed in malignancies and solid tumors. Danusertib (PHA-739358) is a small molecule pan- Aurora kinase inhibitor that also has activity against other cancer-relevant kinases such as Bcr-Abl tyrosine kinase. Other off target effects of danusertib include inhibition of receptor tyrosine kinases such as FGFR-1, and TrkA. Danusertib demonstrates nanomolar IC(50) values for the Aurora kinases all under 100nM and it also demonstrates potent anti-proliferation effects in cell proliferation assays with IC(50) values ranging from 20nM for A2780 cells to 200nM.
- CAS Number:827318-97-8
- Formula:C₂₆H₃₀N₆O₃
- MDL Number:MFCD12024692
- Molecular Weight:474.6 g/mol
- Storage Temperature:Freezer
Specifications
Specification Test Results
- Purity:95%
- Appearance/Color:Solid.
- Solubility:Soluble in DMSO or ethanol.
Specifications
- Pack type:Plastic vial
- Size:5 mg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



