Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Hypericin

Vial, 10 mg
Catalog # 102988-310
Marketsource
Supplier:   Adipogen
 :  AG-CN2-0449-M010
Hypericin
Catalog # 102988-310
Supplier:  Adipogen
Marketsource
CAS Number:  548-04-9
Hypericin

Vial, 10 mg

CAS Number:  548-04-9
Price
$332.31
Log in for availability
Each (10mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

One of the principal active constituents of St. John's Wort (Hypericum perforatum). Hypericin is a photosensitive pigment/chromophore with bright red fluorescence emission (lambdamax: 594nm). Has a high triplet quantum yield and ability to produce considerable amounts of singlet oxygen and other ROS species when photoactivated. Antiviral, anticancer and antidepressant agent. Inactivates enveloped viruses (including HIV). This photosensitizing agent has shown to induce apoptosis and necrosis in cancer cells during photodynamic therapy. Selective inhibitor of protein kinase C (PKC). Has been shown to inhibit other kinases and targets such as EGFR-PTK, PI3K, CKII, MAPK and insulin receptor (IR). Necrosis-avid contrast agent investigated for use in non-invasively targeting necrotic tissues, based on to be defined components in the phospholipid bilayer.

Synonyms:  1,3,4,6,8,13-Hexahydroxy-10,11-dimethylphenanthro[1,10,9,8-opqra]perylene-7,14-dione P-conformer
  • CAS Number:548-04-9
  • Formula:C₃₀H₁₆O₈
  • MDL Number:MFCD00016683
  • Molecular Weight:504.45 g/mol
Specifications

Specifications

  • Pack type:
    Vial
  • Size:
    10 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.