Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BTEC-ALI DIFF MEDIA 100 ML
Catalog # 102970-796
Supplier:
Lifeline Cell Technology
SDS
Expand image
BTEC-ALI DIFF MEDIA 100 ML
Catalog # 102970-796
Supplier:
Lifeline Cell Technology
Supplier Number:
LM-0049
SDS
Related Information
Documentation
SDS
ghs_LM-0049
Documentation