Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
LUNG MICROVASCULAR ENDOTHELIAL CELLS
Catalog # 102970-710
Supplier:
Lifeline Cell Technology
SDS
Expand image
LUNG MICROVASCULAR ENDOTHELIAL CELLS
Catalog # 102970-710
Supplier:
Lifeline Cell Technology
Supplier Number:
FC-0058
SDS
Related Information
Documentation
SDS
ghs_FC-0058
Datasheet
Normal Human Microvascular Endothelial Cells
Documentation