Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
EasyCurve™ Mop System, Contec®

EasyCurve™ Mop System, Contec®

EasyCurve™ Sterile Sealed Edge Mop Head, 38×13×1 cm (15×5×0.375")
Catalog # 10816-298
Supplier:   Contec
Supplier #:  EZSE0150
EasyCurve™ Mop System, Contec®
Catalog # 10816-298
Supplier:  Contec
EasyCurve™ Mop System, Contec®
EasyCurve™ Mop System, Contec®

EasyCurve™ Sterile Sealed Edge Mop Head, 38×13×1 cm (15×5×0.375")

Price
$516.00
Log in for availability
Case of 24
Product Details & Documents

The EasyCurve™ Mop was designed specifically for use in critical and controlled environments. It is now available with a ribbed microfiber face, ensuring smooth and easy mopping with improved particle pick-up and contamination removal.

  • All components are autoclavable; ideal for a wide range of cleaning applications in the most critical sterile environments
  • Lightweight mop head and handle significantly reduces operator fatigue
  • Single-use mop is economical and easy to install without tools
  • Curved mop and pivoting mop frame provide effortless contact with surface
  • Ribbed microfiber face provides better contaminant removal

The combination of the curved mop face and the pivoting connector results in effortless surface contact and outstanding maneuverability. This makes the EasyCurve™ an ideal cleaning tool for ceilings, walls, and floors. All components of the EasyCurve™ Mop are autoclavable. Available in both nonsterile and validated sterile versions.

Specifications
  • Description:
    EasyCurve™ Sterile Sealed Edge Mop Head
  • Size:
    38×13×1 cm (15×5×0.375")
Accessories

Product Information

  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ anodized aluminum telescopic handle
    QuickConnect™ anodized aluminum telescopic handle
    33502-715
    127 - 234 cm (50 - 92")
    Log in for availability
    Each - $204.00

Product Information

  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ stainless steel telescopic handle
    QuickConnect™ stainless steel telescopic handle
    33502-716
    41 - 76 cm (16 - 30")
    Log in for availability
    Each - $385.00

Product Information

  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ anodized aluminum telescopic handle
    QuickConnect™ anodized aluminum telescopic handle
    33502-714
    41 - 76 cm (16 - 30")
    Log in for availability
    Each - $180.00

Product Information

  • Catalog #
    Availability
    Price
  • Stainless Steel Connecting Kit for Multi-Bucket System
    Stainless Steel Connecting Kit for Multi-Bucket System
    89073-986
    Log in for availability
    Each - $97.10

Product Information

  • Catalog #
    Length
    Availability
    Price
  • QuickConnect™ stainless steel telescopic handle
    QuickConnect™ stainless steel telescopic handle
    33502-717
    127 - 234 cm (50 - 92")
    Log in for availability
    Each - $561.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.