Orders LinkContactUsLinkComponent
Interlocking Storage Cabinets, Quantum Storage Systems
Interlocking Storage Cabinets, Quantum Storage Systems
Interlocking cabinet, 8 drawers, 11¾", 11⅜", 11"
Catalog # 10808-754
Supplier #:  QIC-83GY
Interlocking Storage Cabinets, Quantum Storage Systems
Catalog # 10808-754
Interlocking Storage Cabinets, Quantum Storage Systems
Interlocking Storage Cabinets, Quantum Storage Systems
Interlocking cabinet, 8 drawers, 11¾", 11⅜", 11"
Price
$248.94
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Interlocking storage cabinets have the unique ability to interlock horizontally and vertically, allowing customization based on storage needs.
  • Pre-drilled holes on the backside of the cabinet allow for wall mounting
  • Cabinets are made from high impact polystyrene
  • Drawers are removable and will not rust or corrode
  • Drawers have a recessed handle and come with a slotted label holder
Drawers have seven divider slots molded in so the drawer can be divided into multiple compartments. Clear window allows for easy identification of parts. Nine pre-configured storage system comes with drawers, clear windows and two dividers per drawer.
Drawers 11 x2 3/4 x2 1/2" Or 11 x5 5/8 x 2 1/2"
Ordering information: Additional dividers can be purchased separately.
Specifications
  • Depth:
    11⅜"
  • Width:
    11¾"
  • Description:
    Interlocking cabinet
  • Height:
    11"
  • Type:
    8 drawers
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Divider
    Divider
    10808-774
    Log in for availability
    Pack of 1 - $28.17
Product Information
  • Catalog #
    Availability
    Price
  • Divider
    Divider
    10808-776
    Log in for availability
    Pack of 1 - $28.85
Product Information
  • Catalog #
    Availability
    Price
  • Divider
    Divider
    10808-778
    Log in for availability
    Pack of 1 - $30.25
Product Information
  • Catalog #
    Availability
    Price
  • Divider
    Divider
    10808-780
    Log in for availability
    Pack of 1 - $35.22