Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CASPASE-GLO(R) 2 ASSAY 10ML
Catalog # PAG0940
Supplier:
Promega Corporation
SDS
Certificates
Expand image
CASPASE-GLO(R) 2 ASSAY 10ML
Catalog # PAG0940
Supplier:
Promega Corporation
Supplier Number:
G0940
SDS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
SDS
ghs_G0940
Documentation