Orders LinkContactUsLinkComponent
PARtition Wall Systems, Quantum Storage Systems
PARtition Wall Systems, Quantum Storage Systems
Single Sided Starter Unit, White
Catalog # 10805-458
Supplier #:  WS70-SS18HC
PARtition Wall Systems, Quantum Storage Systems
Catalog # 10805-458
PARtition Wall Systems, Quantum Storage Systems
PARtition Wall Systems, Quantum Storage Systems
Single Sided Starter Unit, White
Price
$828.14
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
The wire construction minimzes dirt accumulation and allows air circulation.
  • Available in 11⁷/₈" and 17⁷/₈" width and in 18" and 36" lengths
Baskets have a ½×½" grid pattern to partition products while letting you know when to reorder product and restock items.
Specifications
  • Description:
    Single Sided Starter Unit, White
Accessories
Product Information
  • Catalog #
    Width
    Depth
    Availability
    Price
  • Single Sided
    Single Sided
    10805-742
    24"
    12"
    Log in for availability
    Each - $1,248.48
Product Information
  • Catalog #
    Width
    Depth
    Availability
    Price
  • Single Sided
    Single Sided
    10805-744
    12"
    12"
    Log in for availability
    Each - $1,366.39
Product Information
  • Catalog #
    Width
    Depth
    Availability
    Price
  • Louvered Panel
    Louvered Panel
    10805-752
    12"
    16"
    Log in for availability
    Each - $571.36
Product Information
  • Catalog #
    Width
    Depth
    Availability
    Price
  • Louvered Panel
    Louvered Panel
    10805-754
    24"
    16"
    Log in for availability
    Each - $815.69
Product Information
  • Catalog #
    Availability
    Price
  • Hanging Basket
    Hanging Basket
    10804-580
    Log in for availability
    Each - $195.76
Product Information
  • Catalog #
    Availability
    Price
  • Hanging Basket
    Hanging Basket
    10804-582
    Log in for availability
    Each - $286.47
Product Information
  • Catalog #
    Availability
    Price
  • Adjustable Bracket Pair
    Adjustable Bracket Pair
    10804-388
    Log in for availability
    Each - $32.60
Product Information
  • Catalog #
    Availability
    Price
  • Hang Bar
    Hang Bar
    10804-390
    Log in for availability
    Each - $82.83
Product Information
  • Catalog #
    Availability
    Price
  • Hang Bar
    Hang Bar
    10804-392
    Log in for availability
    Each - $92.77
Product Information
  • Catalog #
    Availability
    Price
  • Hanging Basket
    Hanging Basket
    10804-348
    Log in for availability
    Each - $331.94
Product Information
  • Catalog #
    Availability
    Price
  • Mobile kit with two casters for Single and Double Side Systems
    Mobile kit with two casters for Single and Double Side Systems
    10804-350
    Log in for availability
    Each - $85.94
Product Information
  • Catalog #
    Availability
    Price
  • Horizontal Tape and Reel Holder
    Horizontal Tape and Reel Holder
    10804-362
    Log in for availability
    Each - $43.30
Product Information
  • Catalog #
    Availability
    Price
  • Vertical Tape and Reel Holder
    Vertical Tape and Reel Holder
    10804-364
    Log in for availability
    Each - $41.87
Product Information
  • Catalog #
    Availability
    Price
  • Single Rod
    Single Rod
    10804-366
    Log in for availability
    Each - $17.79
Product Information
  • Catalog #
    Availability
    Price
  • Double Rod
    Double Rod
    10804-368
    Log in for availability
    Each - $28.04
Product Information
  • Catalog #
    Availability
    Price
  • Heavy-Duty
    Heavy-Duty
    10804-370
    Log in for availability
    Each - $26.30
Product Information
  • Catalog #
    Availability
    Price
  • Heavy-Duty
    Heavy-Duty
    10804-372
    Log in for availability
    Each - $28.93
Product Information
  • Catalog #
    Availability
    Price
  • Dust Shelf
    Dust Shelf
    10804-374
    Log in for availability
    Each - $106.63
Product Information
  • Catalog #
    Availability
    Price
  • Dust Shelf
    Dust Shelf
    10804-376
    Log in for availability
    Each - $160.69
Product Information
  • Catalog #
    Availability
    Price
  • Dust Shelf
    Dust Shelf
    10804-378
    Log in for availability
    Each - $222.76
Product Information
  • Catalog #
    Availability
    Price
  • Dust Shelf
    Dust Shelf
    10804-380
    Log in for availability
    Each - $138.45
Product Information
  • Catalog #
    Availability
    Price
  • Dust Shelf
    Dust Shelf
    10804-382
    Log in for availability
    Each - $208.63
Product Information
  • Catalog #
    Availability
    Price
  • Dust Shelf
    Dust Shelf
    10804-384
    Log in for availability
    Each - $278.00
Product Information
  • Catalog #
    Availability
    Price
  • 900 Bracket Pair
    900 Bracket Pair
    10804-386
    Log in for availability
    Each - $19.93
Product Information
  • Catalog #
    Availability
    Price
  • Hanging Basket
    Hanging Basket
    10804-346
    Log in for availability
    Each - $223.62
Product Information
  • Catalog #
    Availability
    Price
  • Louvered Panel
    Louvered Panel
    10804-522
    Log in for availability
    Each - $321.17
Product Information
  • Catalog #
    Availability
    Price
  • Panel Frame
    Panel Frame
    10804-524
    Log in for availability
    Each - $208.89
Product Information
  • Catalog #
    Availability
    Price
  • Panel Frame
    Panel Frame
    10804-526
    Log in for availability
    Each - $229.79
Product Information
  • Catalog #
    Availability
    Price
  • Panel Frame
    Panel Frame
    10804-528
    Log in for availability
    Each - $253.17