REVERSIBLE STRAINER 37UM CS12
Catalog # MSPP-27250
Supplier: Stemcell Technologies
REVERSIBLE STRAINER 37UM CS12
Catalog # MSPP-27250
Supplier: Stemcell Technologies
Supplier Number: 27250
Frequently Bought Together
Acrodisc® PSF Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Whatman™ Uniflo™ Syringe Filters, Whatman products (Cytiva)
Supplier: Cytiva
J.T.Baker®, Syringe Filters
Supplier: Avantor
ANTIBODY CD25 2A3 PE MAB HU 50TEST
Supplier: STEMCELL Technologies
Titan3™ Syringe Filters, 30 mm, Thermo Scientific
Supplier: Thermo Scientific
Diamond® PureFlow™ PES Syringe Filters, Non Sterile, Globe Scientific
Supplier: Globe Scientific
ANTI-HUMAN CD3
Supplier: STEMCELL Technologies
Phage Display Competent Cells, Biosearch Technologies
Supplier: Lucigen
Minisart® SRP Syringe Filters, Sartorius
Supplier: Sartorius
STRAINER REVERSIBLE 37UM SMALL PK50
Supplier: STEMCELL Technologies
Bovine Serum Albumin (BSA)
Supplier: MilliporeSigma