Screw Caps, MicroSolv™
PP, Tan Silicone Rubber / White PTFE, bonded, 9-425 mm, Green, Screw cap
Catalog # 10804-646
Supplier: MicroSolv Technology Corp
Supplier #: 9502S-11-MG
Jump to:
Product Details & Documents
Specifications
- Cap Size:9-425 mm
- Cap type:Screw cap
- Septum:Tan Silicone Rubber / White PTFE, bonded
- Material:PP
- Cap color:Green