Orders LinkContactUsLinkComponent
Conductive Ultra Stack Bins, Quantum Storage Systems
Conductive Ultra Stack Bins, Quantum Storage Systems
Conductive Hang and Stack Bins, 17 1/8 x 14 3/4 x 10 1/4", 18 x 16 1/2 x 11"
Catalog # 10804-248
Supplier #:  QUS270CO
Conductive Ultra Stack Bins, Quantum Storage Systems
Catalog # 10804-248
Conductive Ultra Stack Bins, Quantum Storage Systems
Conductive Ultra Stack Bins, Quantum Storage Systems
Conductive Hang and Stack Bins, 17 1/8 x 14 3/4 x 10 1/4", 18 x 16 1/2 x 11"
Price
$805.01
Log in for availability
Each
Minimum order quantity is 3
Product Details & Documents
Easy access to stored items with hopper front bin and label holder provides fast identification
Protect your electronic components from typical electromagnetic interference, electrostatic or induced fields with these conductive polypropylene bins. The conductive material provides a barrier which these fileds cannot penetrate and prevents the build-up of electrostatic charge. The surface resistivity is less than 105 Ohms-per square.
Reinforced side walls and wide legde allow high stacking on benches or shelving. Bins are permantly conductive and are unaffected by washing with normal cleaners. Be sure all units are properly grounded accoring to your use. Black color.
Ordering information: Conductive Hand and Stack Bins in larger sizes (10804-244, 10804-246, and 10804-248) have a minimum order quantity of 4.
Specifications
  • Description:
    Conductive Hang and Stack Bins
  • Interior dimensions:
    17 1/8 x 14 3/4 x 10 1/4"
  • Ext. dimensions:
    18 x 16 1/2 x 11"
Accessories
Product Information
  • Catalog #
    Fits
    Availability
    Price
  • Conductive Divider for Hang and Stack Bins, Black
    Conductive Divider for Hang and Stack Bins, Black
    10804-564
    QUS255CO bins
    Log in for availability
    Pack of 1 - $162.34
Product Information
  • Catalog #
    Fits
    Availability
    Price
  • Conductive Divider for Hang and Stack Bins, Black
    Conductive Divider for Hang and Stack Bins, Black
    10804-558
    QUS239CO bins
    Log in for availability
    Pack of 1 - $106.70
Product Information
  • Catalog #
    Fits
    Availability
    Price
  • Conductive Divider for Hang and Stack Bins, Black
    Conductive Divider for Hang and Stack Bins, Black
    10804-566
    QUS260CO bins
    Log in for availability
    Pack of 1 - $195.57
Product Information
  • Catalog #
    Fits
    Availability
    Price
  • Conductive Divider for Hang and Stack Bins, Black
    Conductive Divider for Hang and Stack Bins, Black
    10804-562
    QUS250CO bins
    Log in for availability
    Pack of 1 - $129.30
Product Information
  • Catalog #
    Fits
    Availability
    Price
  • Conductive Divider for Hang and Stack Bins, Black
    Conductive Divider for Hang and Stack Bins, Black
    10804-556
    QUS235CO bins
    Log in for availability
    Pack of 1 - $65.01
Product Information
  • Catalog #
    Fits
    Availability
    Price
  • Conductive Divider for Hang and Stack Bins, Black
    Conductive Divider for Hang and Stack Bins, Black
    10804-560
    QUS240CO bins
    Log in for availability
    Pack of 1 - $129.30
Product Information
  • Catalog #
    Fits
    Availability
    Price
  • Conductive Divider for Hang and Stack Bins, Black
    Conductive Divider for Hang and Stack Bins, Black
    10804-252
    QUS210CO bins
    Log in for availability
    Pack of 1 - $33.41
Product Information
  • Catalog #
    Fits
    Availability
    Price
  • Conductive Divider for Hang and Stack Bins, Black
    Conductive Divider for Hang and Stack Bins, Black
    10804-256
    QUS230CO bins
    Log in for availability
    Pack of 1 - $65.01
Product Information
  • Catalog #
    Fits
    Availability
    Price
  • Conductive Divider for Hang and Stack Bins, Black
    Conductive Divider for Hang and Stack Bins, Black
    10804-254
    QUS220CO bins
    Log in for availability
    Pack of 1 - $36.39
Product Information
  • Catalog #
    Fits
    Availability
    Price
  • Conductive Divider for Hang and Stack Bins, Black
    Conductive Divider for Hang and Stack Bins, Black
    10804-568
    QUS270CO bins
    Log in for availability
    Pack of 1 - $201.54