Screw Caps, MicroSolv™
PP, Silicone Rubber / PTFE Pre-Slit, 13 - 425 mm, White, Screw cap
10804-134
: 9504S-30C-W-K
These screw caps are designed for use with 13-425 cap size, 15x45mm vials.
- Cap Size:13 - 425 mm
- Cap type:Screw cap
- Septum:Silicone Rubber / PTFE Pre-Slit
- Material:PP
- Cap color:White